Gene Transcripts

Gene Symbol
LINC01554
Entrez Gene ID
202299
Gene Name
long intergenic non-protein coding RNA 1554
Gene Aliases
C5orf27, FIS
Chromosome Location
Chr. 5: 95852232 - 95860133 on Build GRCh38
UniGene ID
Hs.8373
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026936.1 3 788
GenBank mRNA AK096140.1 - 4 687
AK314737.1 - 3 788
AY168789.1 - 4 667
BC093643.1 - 2 297
BC101577.1 - 2 297

View Full Product Details