Gene Transcripts

Gene Symbol
C11orf45
Entrez Gene ID
219833
Gene Name
chromosome 11 open reading frame 45
Gene Aliases
Chromosome Location
Chr. 11: 128899565 - 128909520 on Build GRCh38
UniGene ID
Hs.689860
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256088.1 NP_001243017.1 2 294
NM_145013.2 NP_659450.1 2 283
NR_045767.1 2 217
XM_011542654.2 XP_011540956.1 2 213
XM_011542655.2 XP_011540957.1 2 175
GenBank mRNA AK125634.1 - 2 191
AK314137.1 - 2 260
BC025756.1 - 2 283 5201867
BX956612.1 - 2 217
DB219954.1 - 2 296

View Full Product Details