Gene Transcripts
- Gene Symbol
- VSTM2A
- Entrez Gene ID
- 222008
- Gene Name
- V-set and transmembrane domain containing 2A
- Gene Aliases
- VSTM2
- Chromosome Location
- Chr. 7: 54542064 - 54571080 on Build GRCh38
- UniGene ID
- Hs.335933
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301009.1 |
NP_001287938.1
|
1, 2 |
473 |
|
NM_001317843.1 |
NP_001304772.1
|
1, 2 |
473 |
|
NM_182546.3 |
NP_872352.2
|
1, 2 |
473 |
|
XM_006715665.3 |
XP_006715728.1
|
1, 2 |
734 |
|
XM_006715666.3 |
XP_006715729.1
|
1, 2 |
734 |
|
XM_017011841.1 |
XP_016867330.1
|
1, 2 |
734 |
|
GenBank mRNA |
BC028404.1 |
- |
1, 2 |
348 |
|
BI754879.1 |
- |
1, 2 |
13 |
|
BI755592.1 |
- |
1, 2 |
473 |
|
BI758069.1 |
- |
1, 2 |
135 |
|
DN991717.1 |
- |
1, 2 |
235 |
|
DN991972.1 |
- |
1, 2 |
473 |
|
View Full Product Details