Gene Transcripts
- Gene Symbol
- SLC39A5
- Entrez Gene ID
- 283375
- Gene Name
- solute carrier family 39 member 5
- Gene Aliases
- LZT-Hs7, MYP24, ZIP5
- Chromosome Location
- Chr. 12: 56230036 - 56237846 on Build GRCh38
- UniGene ID
- Hs.591018
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135195.1 |
NP_001128667.1
|
9, 10 |
1518 |
|
NM_173596.2 |
NP_775867.2
|
11, 12 |
1547 |
|
XM_005268803.1 |
XP_005268860.1
|
9, 10 |
1435 |
|
XM_011538198.1 |
XP_011536500.1
|
9, 10 |
1500 |
|
XM_011538199.1 |
XP_011536501.1
|
11, 12 |
1554 |
|
XM_011538200.1 |
XP_011536502.1
|
10, 11 |
1504 |
|
XM_011538201.1 |
XP_011536503.1
|
10, 11 |
1478 |
|
XM_017019186.1 |
XP_016874675.1
|
11, 12 |
1687 |
|
XM_017019187.1 |
XP_016874676.1
|
9, 10 |
1436 |
|
GenBank mRNA |
AK172768.1 |
- |
9, 10 |
1481 |
|
AK313188.1 |
- |
11, 12 |
1510 |
|
BC027884.1 |
- |
9, 10 |
1392 |
5221329 |
View Full Product Details