Gene Transcripts

Gene Symbol
CATSPER3
Entrez Gene ID
347732
Gene Name
cation channel sperm associated 3
Gene Aliases
CACRC
Chromosome Location
Chr. 5: 134967906 - 135011707 on Build GRCh38
UniGene ID
Hs.631804
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178019.2 NP_821138.1 2 228
GenBank mRNA AF432876.1 - 2 210
AK292427.1 - 2 227
BC101692.1 - 2 188 8069198
BC110384.1 - 2 228 5166215
BI827754.1 - 2 228

View Full Product Details