Gene Transcripts
- Gene Symbol
- RHCE
- Entrez Gene ID
- 6006
- Gene Name
- Rh blood group CcEe antigens
- Gene Aliases
- CD240CE, RH, RH30A, RHC, RHE, RHIXB, RHPI, Rh4, RhIVb(J), RhVI, RhVIII
- Chromosome Location
- Chr. 1: 25362249 - 25430193 on Build GRCh38
- UniGene ID
- Hs.449968
- Species
- Homo sapiens
- Gene Symbol
- RHD
- Entrez Gene ID
- 6007
- Gene Name
- Rh blood group D antigen
- Gene Aliases
- CD240D, DIIIc, RH, RH30, RHCED, RHDVA(TT), RHDel, RHPII, RHXIII, Rh4, RhDCw, RhII, RhK562-II, RhPI
- Chromosome Location
- Chr. 1: 25272393 - 25330445 on Build GRCh38
- UniGene ID
- Hs.449968
- Species
- Homo sapiens
View Full Product Details