Gene Transcripts

Gene Symbol
AKR7A2
Entrez Gene ID
8574
Gene Name
aldo-keto reductase family 7 member A2
Gene Aliases
AFAR, AFAR1, AFB1-AR1, AKR7
Chromosome Location
Chr. 1: 19302708 - 19312146 on Build GRCh38
UniGene ID
Hs.571886
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320979.1 NP_001307908.1 2 430
NM_003689.3 NP_003680.2 2 430
GenBank mRNA AF026947.1 - 2 398
AK225267.1 - 2 401
AK310609.1 - 2 387
BC004111.2 - 2 357 3832140
BC007352.2 - 2 405
BC010852.1 - 2 406 3898226
BC011586.1 - 2 357 4620484
BC012171.1 - 2 405 4642811
BC013996.2 - 2 361 3625337
BG386786.1 - 431
BT007347.1 - 2 321
BX401279.2 - 2 378
EU794591.1 - 2 430
Y16675.1 - 2 392

View Full Product Details