Gene Transcripts

Gene Symbol
RPL14
Entrez Gene ID
9045
Gene Name
ribosomal protein L14
Gene Aliases
CAG-ISL-7, CTG-B33, L14, RL14, hRL14
Chromosome Location
Chr. 3: 40457292 - 40462372 on Build GRCh38
UniGene ID
Hs.446522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034996.2 NP_001030168.1 6 874
NM_003973.4 NP_003964.3 6 810

View Full Product Details