Gene Transcripts
- Gene Symbol
- AKIP1
- Entrez Gene ID
- 56672
- Gene Name
- A-kinase interacting protein 1
- Gene Aliases
- BCA3, C11orf17
- Chromosome Location
- Chr. 11: 8911117 - 8920079 on Build GRCh38
- UniGene ID
- Hs.131180
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001206646.1 |
NP_001193575.1
|
4, 5 |
478 |
|
NM_020642.3 |
NP_065693.2
|
5, 6 |
559 |
|
NR_045417.1 |
|
4, 5 |
741 |
|
NR_045418.1 |
|
3, 4 |
660 |
|
XM_017018011.1 |
XP_016873500.1
|
5, 6 |
579 |
|
GenBank mRNA |
AF493783.1 |
- |
5, 6 |
559 |
|
AF493785.1 |
- |
4, 5 |
478 |
|
AF512007.1 |
- |
4, 5 |
483 |
|
AK075308.1 |
- |
|
678 |
|
BC030996.2 |
- |
4, 5 |
489 |
4609949 |
BM706322.1 |
- |
3, 4 |
514 |
|
View Full Product Details