Gene Transcripts

Gene Symbol
BMP8A
Entrez Gene ID
353500
Gene Name
bone morphogenetic protein 8a
Gene Aliases
Chromosome Location
Chr. 1: 39491626 - 39529869 on Build GRCh38
UniGene ID
Hs.472497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181809.3 NP_861525.2 7 4047
GenBank mRNA AK024413.1 - 1 460
AK127199.1 - 1 1632
BC093743.1 - 1 223
BC112044.1 - 1 223

View Full Product Details