Gene Transcripts
- Gene Symbol
- ASXL3
- Entrez Gene ID
- 80816
- Gene Name
- additional sex combs like 3, transcriptional regulator
- Gene Aliases
- BRPS, KIAA1713
- Chromosome Location
- Chr. 18: 33578577 - 33751195 on Build GRCh38
- UniGene ID
- Hs.464876
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030632.2 |
NP_085135.1
|
9, 10 |
1026 |
|
XM_005258356.1 |
XP_005258413.1
|
10, 11 |
999 |
|
XM_011526205.2 |
XP_011524507.1
|
10, 11 |
7789 |
|
XM_011526206.2 |
XP_011524508.1
|
10, 11 |
4554 |
|
XM_011526209.1 |
XP_011524511.1
|
11, 12 |
1183 |
|
XM_011526210.1 |
XP_011524512.1
|
11, 12 |
1240 |
|
XM_011526212.1 |
XP_011524514.1
|
10, 11 |
1144 |
|
XM_011526213.1 |
XP_011524515.1
|
10, 11 |
1237 |
|
XM_017026012.1 |
XP_016881501.1
|
10, 11 |
4401 |
|
XM_017026013.1 |
XP_016881502.1
|
11, 12 |
4763 |
|
XM_017026014.1 |
XP_016881503.1
|
11, 12 |
1200 |
|
GenBank mRNA |
AK131454.1 |
- |
11, 12 |
1069 |
|
View Full Product Details