Gene Transcripts

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
Chromosome Location
Chr. 7: 978485 - 1138291 on Build GRCh38
UniGene ID
Hs.653258
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134395.1 NP_001127867.1 5 630
NM_001134396.1 NP_001127868.1 5 611
NM_001318252.1 NP_001305181.1 5 1514
NM_032350.5 NP_115726.1 5 640
NR_134537.1 4 616
GenBank mRNA AF495721.1 - 5 1470
BC006224.2 - 5 640 3941780
BM676072.1 - 1 668

View Full Product Details