Gene Transcripts

Gene Symbol
ATPIF1
Entrez Gene ID
93974
Gene Name
ATPase inhibitory factor 1
Gene Aliases
ATPI, ATPIP, IP
Chromosome Location
Chr. 1: 28236091 - 28238105 on Build GRCh38
UniGene ID
Hs.744914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016311.4 NP_057395.1 3 348
NM_178190.2 NP_835497.1 3 424
NM_178191.2 NP_835498.1 2 1732
GenBank mRNA AB029042.1 - 3 276
AF114836.1 - 434
AK000934.1 - 2 1669
AK316600.1 - 3 335
AL050386.1 - 2 1662
AL583540.3 - 3 347
BC004955.1 - 3 231 3625379
BC009677.1 - 3 306 3877506
BG036978.1 - 3 396
BG337531.1 - 3 304
BP271519.1 - 3 344
BT009849.1 - 3 255
CR457097.1 - 3 255

View Full Product Details