Gene Transcripts

Gene Symbol
C19orf25
Entrez Gene ID
148223
Gene Name
chromosome 19 open reading frame 25
Gene Aliases
Chromosome Location
Chr. 19: 1473201 - 1479552 on Build GRCh38
UniGene ID
Hs.532840
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152482.2 NP_689695.2 3 222
XM_005259506.3 XP_005259563.1 3 238
XM_006722653.3 XP_006722716.1 3 197
XM_017026374.1 XP_016881863.1 3 545
GenBank mRNA AK056004.1 - 1 1250
AK075267.1 - 3 205
AK093985.1 - 3 187
AK297944.1 - 3 329
AL122089.1 - 3 193
AY007120.1 - 2 158
BC018441.1 - 3 192 4471390
BP210473.1 - 3 222

View Full Product Details