Gene Transcripts

Gene Symbol
FAM120AOS
Entrez Gene ID
158293
Gene Name
family with sequence similarity 120A opposite strand
Gene Aliases
C9orf10OS
Chromosome Location
Chr. 9: 93446500 - 93453592 on Build GRCh38
UniGene ID
Hs.350364
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198841.2 NP_942138.2 1 918
NR_136231.1 1 918
GenBank mRNA AK056096.1 - 1 918

View Full Product Details