Gene Transcripts
- Gene Symbol
- AMZ2P1
- Entrez Gene ID
- 201283
- Gene Name
- archaelysin family metallopeptidase 2 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 17: 64966550 - 64975585 on Build GRCh38
- UniGene ID
- Hs.396447
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026903.1 |
|
5 |
2967 |
|
GenBank mRNA |
AK056627.1 |
- |
5 |
2950 |
|
View Full Product Details