Gene Transcripts

Gene Symbol
C12orf71
Entrez Gene ID
728858
Gene Name
chromosome 12 open reading frame 71
Gene Aliases
Chromosome Location
Chr. 12: 27081057 - 27084548 on Build GRCh38
UniGene ID
Hs.334003
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080406.1 NP_001073875.1 1 426
XM_011520847.2 XP_011519149.1 2 968

View Full Product Details