Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011538259.2 | XP_011536561.1 | 1 | 564 | |
XM_011538263.2 | XP_011536565.1 | 1 | 564 | ||
XM_011538264.2 | XP_011536566.1 | 1 | 563 | ||
XM_011538265.2 | XP_011536567.1 | 1 | 566 |