Gene Transcripts

Gene Symbol
LINC01551
Entrez Gene ID
387978
Gene Name
long intergenic non-protein coding RNA 1551
Gene Aliases
C14orf23, c14_5148
Chromosome Location
Chr. 14: 28772704 - 28794794 on Build GRCh38
UniGene ID
Hs.633254
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026732.1 2, 3 219
GenBank mRNA BM839073.1 - 2, 3 138

View Full Product Details