Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162491.1 | NP_001155963.1 | 5, 6 | 600 | |
NR_103708.1 | 5, 6 | 600 | |||
NR_103709.1 | 5, 6 | 600 | |||
XM_011530950.2 | XP_011529252.1 | 5, 6 | 1297 | ||
GenBank mRNA | AK301813.1 | - | 5, 6 | 600 | |
BC140808.1 | - | 5, 6 | 674 | 9021676 | |
BC144603.1 | - | 5, 6 | 674 | 9053134 | |
CD300373.1 | - | 2, 3 | 324 |