Gene Transcripts

Gene Symbol
ABCB5
Entrez Gene ID
340273
Gene Name
ATP binding cassette subfamily B member 5
Gene Aliases
ABCB5alpha, ABCB5beta, EST422562
Chromosome Location
Chr. 7: 20615622 - 20757014 on Build GRCh38
UniGene ID
Hs.404102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163941.1 NP_001157413.1 9 1167
GenBank mRNA AB353947.1 - 9 1167
AY230001.1 - 9 1060

View Full Product Details