Gene Transcripts

Gene Symbol
C22orf39
Entrez Gene ID
128977
Gene Name
chromosome 22 open reading frame 39
Gene Aliases
Chromosome Location
Chr. 22: 19440886 - 19448232 on Build GRCh38
UniGene ID
Hs.659565
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166242.1 NP_001159714.1 1 148
NM_173793.4 NP_776154.3 1 148
GenBank mRNA AK290073.1 - 1 121
BX337505.2 - 1 148

View Full Product Details