Gene Transcripts
- Gene Symbol
- SNORA6
- Entrez Gene ID
- 574040
- Gene Name
- small nucleolar RNA, H/ACA box 6
- Gene Aliases
- ACA6
- Chromosome Location
- Chr. 3: 39408391 - 39408539 on Build GRCh38
- UniGene ID
- Hs.745521
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002325.1 |
|
1 |
12 |
|
GenBank mRNA |
AF375472.1 |
- |
1 |
12 |
|
AJ609429.1 |
- |
1 |
12 |
|
View Full Product Details