Gene Transcripts
- Gene Symbol
- ABLIM2
- Entrez Gene ID
- 84448
- Gene Name
- actin binding LIM protein family member 2
- Gene Aliases
- Chromosome Location
- Chr. 4: 7965310 - 8158832 on Build GRCh38
- UniGene ID
- Hs.233404
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130044.1 |
- |
1 |
2402 |
|
View Full Product Details