Gene Transcripts

Gene Symbol
BSPH1
Entrez Gene ID
100131137
Gene Name
binder of sperm protein homolog 1
Gene Aliases
BSP1, ELSPBP2
Chromosome Location
Chr. 19: 47967258 - 47992170 on Build GRCh38
UniGene ID
Hs.651072
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128326.2 NP_001121798.1 5 396
XM_017026118.1 XP_016881607.1 5 396
GenBank mRNA DQ227497.1 - 5 397

View Full Product Details