Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001122957.2 | NP_001116429.1 | 11 | 1314 | |
NM_001271926.1 | NP_001258855.1 | 10 | 1224 | ||
NM_005881.3 | NP_005872.2 | 11 | 1314 | ||
XM_017022859.1 | XP_016878348.1 | 11 | 1227 | ||
GenBank mRNA | AF026548.1 | - | 11 | 1224 | |
AK130145.1 | - | 11 | 1194 | ||
BC007363.2 | - | 11 | 1197 | 3630050 | |
BC009872.2 | - | 10 | 1087 | 3944953 | |
CR542093.1 | - | 10 | 951 |