Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127325.1 | NP_001120797.1 | 3 | 344 | |
NM_006341.3 | NP_006332.3 | 3 | 311 | ||
XM_011540507.1 | XP_011538809.1 | 3 | 172 | ||
GenBank mRNA | AF072933.1 | - | 3 | 155 | |
AF080398.1 | - | 2 | 140 | ||
AF139365.1 | - | 3 | 234 | ||
AF157482.1 | - | 3 | 311 | ||
AK027327.1 | - | 3 | 224 | ||
AK094316.1 | - | 5 | 1053 | ||
BC015244.1 | - | 2 | 125 | 3916742 | |
BI517654.1 | - | 3 | 344 |