Gene Transcripts

Gene Symbol
LILRA1
Entrez Gene ID
11024
Gene Name
leukocyte immunoglobulin like receptor A1
Gene Aliases
CD85I, LIR-6, LIR6
Chromosome Location
Chr. 19: 54593576 - 54602381 on Build GRCh38
UniGene ID
Hs.710507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278318.1 NP_001265247.1 2, 3 281
NM_001278319.1 NP_001265248.1 1, 2 75
NM_006863.3 NP_006854.1 2, 3 281
GenBank mRNA AF025530.1 - 2, 3 209
BC093708.1 - 1, 2 55 7939553
BC101514.1 - 1, 2 55 8069020
EU915602.1 - 1, 2 44

View Full Product Details