Gene Transcripts
- Gene Symbol
- CASC4
- Entrez Gene ID
- 113201
- Gene Name
- cancer susceptibility candidate 4
- Gene Aliases
- H63
- Chromosome Location
- Chr. 15: 44288616 - 44415761 on Build GRCh38
- UniGene ID
- Hs.512867
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138423.3 |
NP_612432.2
|
1, 2 |
643 |
|
NM_177974.2 |
NP_816929.1
|
1, 2 |
643 |
|
XM_017021880.1 |
XP_016877369.1
|
1, 2 |
738 |
|
GenBank mRNA |
AK291026.1 |
- |
1, 2 |
400 |
|
AK298564.1 |
- |
2, 3 |
358 |
|
AL832333.1 |
- |
1, 2 |
415 |
|
AL832840.1 |
- |
1, 2 |
85 |
|
AY358086.1 |
- |
1, 2 |
594 |
|
BC012124.1 |
- |
1, 2 |
623 |
|
BC063480.1 |
- |
1, 2 |
597 |
5218924 |
View Full Product Details