Gene Transcripts

Gene Symbol
C22orf15
Entrez Gene ID
150248
Gene Name
chromosome 22 open reading frame 15
Gene Aliases
N27C7-3
Chromosome Location
Chr. 22: 23762522 - 23765863 on Build GRCh38
UniGene ID
Hs.116254
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182520.2 NP_872326.2 3 487
XM_011529907.2 XP_011528209.1 2 1278
XM_011529908.2 XP_011528210.2 3 984
XM_017028602.1 XP_016884091.1 2 1278
XM_017028603.1 XP_016884092.1 2 1257
XM_017028604.1 XP_016884093.1 2 1278
XM_017028605.1 XP_016884094.1 2 1278
XM_017028606.1 XP_016884095.1 2 1278
XM_017028607.1 XP_016884096.1 2 1278
XM_017028608.1 XP_016884097.1 2 1278
XM_017028609.1 XP_016884098.1 3 1007
XM_017028610.1 XP_016884099.1 3 985
XM_017028611.1 XP_016884100.1 2 1278
XM_017028612.1 XP_016884101.1 3 983
XM_017028613.1 XP_016884102.1 2 1278
XM_017028614.1 XP_016884103.1 2 1278
XM_017028615.1 XP_016884104.1 3 984
XM_017028616.1 XP_016884105.1 3 985
XM_017028617.1 XP_016884106.1 3 986
XM_017028618.1 XP_016884107.1 3 508
GenBank mRNA AK093880.1 - 3 508
BU584544.1 - 3 483
BU739625.1 - 4 389
CR456371.1 - 3 285
DB200363.1 - 3 508

View Full Product Details