Gene Transcripts

Gene Symbol
SRFBP1
Entrez Gene ID
153443
Gene Name
serum response factor binding protein 1
Gene Aliases
BUD22, P49, Rlb1, STRAP, p49/STRAP
Chromosome Location
Chr. 5: 121961917 - 122075624 on Build GRCh38
UniGene ID
Hs.107622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152546.2 NP_689759.2 5 375
XM_017009111.1 XP_016864600.1 5 419
GenBank mRNA AK058015.1 - 5 375
AY611630.2 - 5 303
BC031222.1 - 5 346 5297296

View Full Product Details