Gene Transcripts
- Gene Symbol
- DMXL2
- Entrez Gene ID
- 23312
- Gene Name
- Dmx like 2
- Gene Aliases
- PEPNS, RC3
- Chromosome Location
- Chr. 15: 51447724 - 51622833 on Build GRCh38
- UniGene ID
- Hs.511386
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001174116.1 |
NP_001167587.1
|
36, 37 |
8493 |
|
NM_001174117.1 |
NP_001167588.1
|
34, 35 |
6582 |
|
NM_015263.3 |
NP_056078.2
|
36, 37 |
8490 |
|
XM_005254255.1 |
XP_005254312.1
|
37, 38 |
8619 |
|
XM_005254256.1 |
XP_005254313.1
|
37, 38 |
8616 |
|
XM_017022034.1 |
XP_016877523.1
|
37, 38 |
8292 |
|
GenBank mRNA |
AB020663.2 |
- |
24, 25 |
5870 |
|
AF389880.1 |
- |
36, 37 |
8472 |
|
BC016032.1 |
- |
14, 15 |
2705 |
|
BC022050.1 |
- |
7, 8 |
810 |
|
BC140781.1 |
- |
34, 35 |
6447 |
9021649 |
BC144539.1 |
- |
36, 37 |
8358 |
9053066 |
View Full Product Details