Gene Transcripts

Gene Symbol
APPL1
Entrez Gene ID
26060
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
Gene Aliases
APPL, DIP13alpha, MODY14
Chromosome Location
Chr. 3: 57227737 - 57273471 on Build GRCh38
UniGene ID
Hs.476415
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012096.2 NP_036228.1 6 547
XM_011533583.2 XP_011531885.1 7 600
GenBank mRNA AB037849.2 - 6 400
AF169797.1 - 6 458
AF424738.1 - 6 456
AK299008.1 - 7 589
AK310716.1 - 6 495
BC028599.2 - 6 547 4829430

View Full Product Details