Gene Transcripts

Gene Symbol
AQP4
Entrez Gene ID
361
Gene Name
aquaporin 4
Gene Aliases
MIWC, WCH4
Chromosome Location
Chr. 18: 26852038 - 26865844 on Build GRCh38
UniGene ID
Hs.315369
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317384.1 NP_001304313.1 3 629
NM_001317387.1 NP_001304316.1 3 629
NM_001650.5 NP_001641.1 3 629
NM_004028.4 NP_004019.1 2 756
XM_011525942.2 XP_011524244.1 3 671
GenBank mRNA AB128929.1 - 4 159
AK222684.1 - 3 541
AK295069.1 - 4 440
BC022286.1 - 3 494 4717755
BI596912.1 - 3 529
BI667387.1 - 3 547
D63412.1 - 3 494
HQ901095.1 - 3 603
KF055862.1 - 3 474
U34845.1 - 2 692
U63622.1 - 3 513
U63623.1 - 2 417

View Full Product Details