Gene Transcripts

Gene Symbol
SERTM1
Entrez Gene ID
400120
Gene Name
serine rich and transmembrane domain containing 1
Gene Aliases
C13orf36
Chromosome Location
Chr. 13: 36673912 - 36697839 on Build GRCh38
UniGene ID
Hs.422375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203451.2 NP_982276.2 2 550
GenBank mRNA AK294236.1 - 2 442
BC036540.2 - 2 550 5260372
BI544475.1 - 2 447

View Full Product Details