Gene Transcripts
- Gene Symbol
- MYL4
- Entrez Gene ID
- 4635
- Gene Name
- myosin light chain 4
- Gene Aliases
- ALC1, AMLC, GT1, PRO1957
- Chromosome Location
- Chr. 17: 47209017 - 47227650 on Build GRCh38
- UniGene ID
- Hs.463300
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001002841.1 |
NP_001002841.1
|
4, 5 |
439 |
|
NM_002476.2 |
NP_002467.1
|
3, 4 |
386 |
|
XM_005257391.4 |
XP_005257448.1
|
4, 5 |
366 |
|
XM_011524839.2 |
XP_011523141.2
|
4, 5 |
526 |
|
XM_017024683.1 |
XP_016880172.1
|
4, 5 |
513 |
|
XM_017024684.1 |
XP_016880173.1
|
3, 4 |
530 |
|
GenBank mRNA |
AF116676.2 |
- |
|
738 |
|
BC030228.1 |
- |
3, 4 |
375 |
5226518 |
BU658678.1 |
- |
4, 5 |
445 |
|
M24121.1 |
- |
3, 4 |
373 |
|
M36172.1 |
- |
3, 4 |
367 |
|
X13955.1 |
- |
3, 4 |
367 |
|
View Full Product Details