Gene Transcripts

Gene Symbol
ANGPTL4
Entrez Gene ID
51129
Gene Name
angiopoietin like 4
Gene Aliases
ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158
Chromosome Location
Chr. 19: 8364127 - 8374375 on Build GRCh38
UniGene ID
Hs.9613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039667.2 NP_001034756.1 2 540
NM_139314.2 NP_647475.1 2 540
NR_104213.1 2 540
XM_005272484.3 XP_005272541.1 2 538
XM_005272485.3 XP_005272542.1 2 538
GenBank mRNA AB056477.1 - 2 514
AF153606.1 - 2 495
AF169312.1 - 2 409
AF202636.1 - 2 517
AK222489.1 - 2 512
AK222524.1 - 2 510
AK303269.1 - 3 441
AY358275.1 - 2 508
BC023647.2 - 2 484 5088323
BG424907.1 - 2 539

View Full Product Details