Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001283035.1 | NP_001269964.1 | 9 | 892 | |
NM_001283036.1 | NP_001269965.1 | 8 | 799 | ||
NM_001283037.1 | NP_001269966.1 | 7 | 747 | ||
NM_016407.4 | NP_057491.2 | 8 | 802 | ||
XM_017027872.1 | XP_016883361.1 | 11 | 995 | ||
GenBank mRNA | AF117231.1 | - | 8 | 750 | |
AF161513.1 | - | 8 | 759 | ||
AF161518.1 | - | 8 | 728 | ||
AF212244.1 | - | 8 | 739 | ||
AK000481.1 | - | 8 | 732 | ||
AK000586.1 | - | 7 | 695 | ||
AK026118.1 | - | 8 | 730 | ||
AK225135.1 | - | 8 | 717 | ||
AK293662.1 | - | 8 | 651 | ||
AK302029.1 | - | 9 | 835 | ||
AK314563.1 | - | 8 | 757 | ||
BC003359.1 | - | 8 | 729 | 2900332 | |
BU537939.1 | - | 8 | 713 | ||
CR983692.1 | - | 7 | 658 |