Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242729.1 | NP_001229658.1 | 3, 4 | 649 | |
NM_017700.1 | NP_060170.1 | 3, 4 | 649 | ||
XM_011532050.2 | XP_011530352.1 | 2, 3 | 198 | ||
XM_011532051.2 | XP_011530353.1 | 3, 4 | 664 | ||
GenBank mRNA | AK000191.1 | - | 3, 4 | 649 | |
BC105695.1 | - | 3, 4 | 637 | 40023595 |