Gene Transcripts

Gene Symbol
LGR4
Entrez Gene ID
55366
Gene Name
leucine rich repeat containing G protein-coupled receptor 4
Gene Aliases
BNMD17, GPR48
Chromosome Location
Chr. 11: 27365961 - 27472787 on Build GRCh38
UniGene ID
Hs.502176
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018490.2 NP_060960.2 5, 6 1097
GenBank mRNA AB209743.1 - 5, 6 1016
AF257182.1 - 5, 6 1056
BC033039.2 - 4, 5 1013 5260923

View Full Product Details