Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000953.2 | NP_000944.1 | 1, 2 | 949 | |
XM_005267891.3 | XP_005267948.1 | 1, 2 | 1406 | ||
GenBank mRNA | BC040968.1 | - | 1, 2 | 949 | |
EF577397.1 | - | 1, 2 | 847 | ||
U31099.1 | - | 1, 2 | 203 |