Gene Transcripts
- Gene Symbol
- MOK
- Entrez Gene ID
- 5891
- Gene Name
- MOK protein kinase
- Gene Aliases
- RAGE, RAGE-1, RAGE1, STK30
- Chromosome Location
- Chr. 14: 102224450 - 102305200 on Build GRCh38
- UniGene ID
- Hs.104119
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014226.2 |
NP_055041.1
|
2, 3 |
355 |
|
XM_006720225.2 |
XP_006720288.1
|
2, 3 |
355 |
|
XM_006720226.2 |
XP_006720289.1
|
2, 3 |
355 |
|
XM_011537052.1 |
XP_011535354.1
|
2, 3 |
355 |
|
XM_017021549.1 |
XP_016877038.1
|
2, 3 |
355 |
|
XM_017021555.1 |
XP_016877044.1
|
2, 3 |
355 |
|
XM_017021556.1 |
XP_016877045.1
|
2, 3 |
355 |
|
GenBank mRNA |
AB022694.1 |
- |
2, 3 |
349 |
|
AK312778.1 |
- |
2, 3 |
346 |
|
BC026069.1 |
- |
2, 3 |
345 |
4838296 |
BC053536.1 |
- |
2, 3 |
331 |
5175851 |
DB037795.1 |
- |
2, 3 |
355 |
|
View Full Product Details