Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001011658.3 | NP_001011658.1 | 4 | 428 | |
NM_001128835.2 | NP_001122307.2 | 4 | 422 | ||
NM_014563.5 | NP_055378.1 | 3 | 286 | ||
XM_011545565.1 | XP_011543867.1 | 5 | 496 | ||
XM_011545566.2 | XP_011543868.1 | 4 | 352 | ||
GenBank mRNA | AK098253.1 | - | 2 | 2435 | |
AK310542.1 | - | 4 | 332 | ||
AK313736.1 | - | 4 | 368 | ||
BC016915.1 | - | 3 | 249 | 4432196 | |
BC052618.1 | - | 3 | 249 | 6581382 | |
BI821518.1 | - | 3 | 260 | ||
CB962763.1 | - | 3 | 271 | ||
CD698535.1 | - | 4 | 382 | ||
DA344866.1 | - | 3 | 286 | ||
DA350753.1 | - | 4 | 416 | ||
DA542477.1 | - | 4 | 362 |