Gene Transcripts

Gene Symbol
ATP13A3
Entrez Gene ID
79572
Gene Name
ATPase 13A3
Gene Aliases
AFURS1
Chromosome Location
Chr. 3: 194402674 - 194487006 on Build GRCh38
UniGene ID
Hs.529609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024524.3 NP_078800.3 12 1676
XM_005269357.3 XP_005269414.1 12 1697
XM_011513120.2 XP_011511422.1 13 1798
XM_011513121.1 XP_011511423.1 12 1676
XM_011513122.1 XP_011511424.1 12 1370
XM_011513123.2 XP_011511425.1 13 1803
XM_011513124.2 XP_011511426.1 13 1803
GenBank mRNA AJ306929.1 - 3 2200
AK024639.1 - 1 73
AK225064.1 - 1 73
DR005713.1 - 5 557

View Full Product Details