Gene Transcripts

Gene Symbol
PHF6
Entrez Gene ID
84295
Gene Name
PHD finger protein 6
Gene Aliases
BFLS, BORJ, CENP-31
Chromosome Location
Chr. X: 134373312 - 134428792 on Build GRCh38
UniGene ID
Hs.356501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015877.1 NP_001015877.1 2, 3 331
NM_032335.3 NP_115711.2 2, 3 331
NM_032458.2 NP_115834.1 2, 3 331
GenBank mRNA AB058726.1 - 2, 3 217
AK290095.1 - 2, 3 302
AL832725.1 - 2, 3 324
AY157622.1 - 2, 3 331
BC005994.1 - 2, 3 270 4064169
CD685336.1 - 2, 3 339
DR000282.1 - 2, 3 366

View Full Product Details