Gene Transcripts

Gene Symbol
PHF6
Entrez Gene ID
84295
Gene Name
PHD finger protein 6
Gene Aliases
BFLS, BORJ, CENP-31
Chromosome Location
Chr. X: 134373312 - 134428792 on Build GRCh38
UniGene ID
Hs.356501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015877.1 NP_001015877.1 4 523
NM_032335.3 NP_115711.2 4 523
NM_032458.2 NP_115834.1 4 523
GenBank mRNA AB058726.1 - 4 409
AK290095.1 - 4 494
AK303369.1 - 3 386
AL832725.1 - 4 516
AY157622.1 - 4 523
BC005994.1 - 4 462 4064169
CD685336.1 - 4 531
DR000282.1 - 4 558

View Full Product Details