Gene Transcripts
- Gene Symbol
- MKNK1
- Entrez Gene ID
- 8569
- Gene Name
- MAP kinase interacting serine/threonine kinase 1
- Gene Aliases
- MNK1
- Chromosome Location
- Chr. 1: 46557407 - 46607140 on Build GRCh38
- UniGene ID
- Hs.371594
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135553.2 |
NP_001129025.1
|
6, 7 |
629 |
|
NM_003684.5 |
NP_003675.2
|
6, 7 |
629 |
|
NM_198973.3 |
NP_945324.1
|
6, 7 |
587 |
|
XM_006711000.1 |
XP_006711063.1
|
6, 7 |
582 |
|
XM_006711001.2 |
XP_006711064.1
|
7, 8 |
832 |
|
XM_006711002.2 |
XP_006711065.1
|
6, 7 |
6057 |
|
XM_006711003.1 |
XP_006711066.1
|
7, 8 |
643 |
|
XM_006711006.3 |
XP_006711069.1
|
5, 6 |
3372 |
|
XM_006711007.1 |
XP_006711070.1
|
5, 6 |
474 |
|
XM_006711009.3 |
XP_006711072.1
|
5, 6 |
5959 |
|
XM_017002652.1 |
XP_016858141.1
|
6, 7 |
6305 |
|
XM_017002653.1 |
XP_016858142.1
|
7, 8 |
930 |
|
XM_017002654.1 |
XP_016858143.1
|
7, 8 |
3782 |
|
XM_017002655.1 |
XP_016858144.1
|
7, 8 |
6403 |
|
XM_017002656.1 |
XP_016858145.1
|
7, 8 |
6202 |
|
XM_017002657.1 |
XP_016858146.1
|
6, 7 |
6104 |
|
XM_017002658.1 |
XP_016858147.1
|
6, 7 |
6305 |
|
XM_017002659.1 |
XP_016858148.1
|
6, 7 |
601 |
|
XM_017002660.1 |
XP_016858149.1
|
6, 7 |
6057 |
|
XM_017002661.1 |
XP_016858150.1
|
7, 8 |
822 |
|
XM_017002662.1 |
XP_016858151.1
|
7, 8 |
643 |
|
XM_017002663.1 |
XP_016858152.1
|
7, 8 |
6403 |
|
XM_017002665.1 |
XP_016858154.1
|
5, 6 |
474 |
|
XM_017002668.1 |
XP_016858157.1
|
6, 7 |
6305 |
|
GenBank mRNA |
AB000409.1 |
- |
6, 7 |
572 |
|
AB451300.1 |
- |
5, 6 |
385 |
|
AB451432.1 |
- |
5, 6 |
385 |
|
AK096423.1 |
- |
7, 8 |
674 |
|
AK290456.1 |
- |
6, 7 |
549 |
|
AK310566.1 |
- |
5, 6 |
474 |
|
AY355461.1 |
- |
5, 6 |
385 |
|
BC002755.2 |
- |
6, 7 |
549 |
3629765 |
BG722049.1 |
- |
6, 7 |
592 |
|
View Full Product Details