Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308025.1 | NP_001294954.1 | 18 | 3074 | |
NM_152259.3 | NP_689472.3 | 18 | 3077 | ||
GenBank mRNA | AK123612.1 | - | 11 | 1177 | |
BX538335.1 | - | 5 | 378 | ||
GQ227787.1 | - | 18 | 3074 |