Gene Transcripts
- Gene Symbol
- UBXN11
- Entrez Gene ID
- 91544
- Gene Name
- UBX domain protein 11
- Gene Aliases
- COA-1, PP2243, SOC, SOCI, UBXD5
- Chromosome Location
- Chr. 1: 26282282 - 26318265 on Build GRCh38
- UniGene ID
- Hs.145061
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077262.1 |
NP_001070730.1
|
2, 3 |
228 |
|
NM_145345.2 |
NP_663320.2
|
3, 4 |
447 |
|
NM_183008.2 |
NP_892120.2
|
3, 4 |
353 |
|
GenBank mRNA |
AF520989.1 |
- |
2, 3 |
156 |
|
AF521017.1 |
- |
2, 3 |
157 |
|
AK093258.1 |
- |
3, 4 |
297 |
|
AK094198.1 |
- |
2, 3 |
160 |
|
AK097368.1 |
- |
3, 4 |
545 |
|
AK291503.1 |
- |
2, 3 |
152 |
|
BC038106.1 |
- |
2, 3 |
165 |
5269682 |
BX647774.1 |
- |
2, 3 |
161 |
|
BX648412.1 |
- |
2, 3 |
161 |
|
CR749413.1 |
- |
2, 3 |
159 |
|
DA421971.1 |
- |
2, 3 |
160 |
|
DA989799.1 |
- |
2, 3 |
261 |
|
View Full Product Details