Gene Transcripts

Gene Symbol
TEX30
Entrez Gene ID
93081
Gene Name
testis expressed 30
Gene Aliases
C13orf27
Chromosome Location
Chr. 13: 102765888 - 102773821 on Build GRCh38
UniGene ID
Hs.398111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286776.1 NP_001273705.1 4 444
NM_138779.4 NP_620134.3 5 519
XM_005254097.2 XP_005254154.1 5 573
XM_011521135.2 XP_011519437.1 5 1160
XM_011521136.2 XP_011519438.1 3 289
XM_017020852.1 XP_016876341.1 5 1504
GenBank mRNA AF070559.1 - 3 1033
AK314995.1 - 5 448
BC015148.2 - 5 490 4825712
BC048011.1 - 2 5551
BI559423.1 - 4 417
BP363423.1 - 4 422
HY045638.1 - 4 444

View Full Product Details